missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanC lantibiotic synthetase component C-like 1 (bacterial), Mouse, Polyclonal Antibody, Abnova™
Description
This gene encodes a loosely associated peripheral membrane protein related to the LanC family of bacterial membrane-associated proteins involved in the biosynthesis of antimicrobial peptides. This protein may play a role as a peptide-modifying enzyme component in eukaryotic cells. Previously considered a member of the G-protein-coupled receptor superfamily, this protein is now in the LanC family. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Sequence: MAQRAFPNPYADYNKSLAEGYFDAAGRLTPEFSQRLTNKIRELLQQMERGLKSADPRD
Specifications
Specifications
| Antigen | LanC lantibiotic synthetase component C-like 1 (bacterial) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant LANCL1. |
| Formulation | 50% glycerol |
| Gene | LANCL1 |
| Gene Accession No. | NM_006055 |
| Gene Alias | GPR69A/p40 |
| Gene Symbols | LANCL1 |
| Show More |