missing translation for 'onlineSavingsMsg'
Learn More

Lactotransferrin Rabbit anti-Human, Mouse, Rat, Hamster, Monkey, Polyclonal, MilliporeSigma™

Catalog No. 07685MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07685MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07685MI Supplier MilliporeSigma Supplier No. 07685

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Anti-Lactotransferrin Antibody is an antibody against Lactotransferrin for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin).

Specifications

Antigen Lactotransferrin
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols LTF; lactotransferrin; HLF2; GIG12; lactoferrin; LF; Lactoferrin
Host Species Rabbit
Immunogen Full length native mouse Lactotransferrin/Lactoferrin
Purification Method Serum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002343.2
Target Species Hamster, Human, Monkey, Mouse, Rat
Content And Storage 2-8°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only