missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L.pneumophila Serogroup 1 Mouse anti-Human, Clone: Lp1 MAB 3, MilliporeSigma™
Description
This Anti-L. pneumophila Serogroup 1 Antibody, clone Lp1 3 is validated for use in IF for the detection of L. pneumophila Serogroup 1.
Specifications
Specifications
| Antigen | L. pneumophila Serogroup 1 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | Lp1 MAB 3 |
| Conjugate | Unconjugated |
| Formulation | Liquid ascites fluid containing no preservatives. |
| Host Species | Mouse |
| Immunogen | Bacterial antigenic subgroup Knoxville 1 |
| Purification Method | Ascites |
| Quantity | 100 μL |
| Show More |
For Research Use Only