Learn More
v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog, Mouse, Clone: 4F3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant KRAS.
Supplier: Abnova Corporation H00003845M02
Description
This gene, a Kirsten ras oncogene homolog from the mammalian ras gene family, encodes a protein that is a member of the small GTPase superfamily. A single amino acid substitution is responsible for an activating mutation. The transforming protein that results is implicated in various malignancies, including lung adenocarcinoma, mucinous adenoma, ductal carcinoma of the pancreas and colorectal carcinoma. Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region. [provided by RefSeq
Sequence: KSALTIQLIQNHFVDEYDPTIEDSYRKQVVIDGETCLLDILDTAGQEEYSAMRDQYMRTGEGFLCVFAINNTKSFEDIHHYREQIKRVKDSEDVPMVLVGNKCDLPSRTVSpecifications
| v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004985 | |
| KRAS | |
| KRAS (NP_004976, 16 a.a. ∼ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4F3 | |
| Mouse monoclonal antibody raised against a partial recombinant KRAS. | |
| KRAS | |
| C-K-RAS/K-RAS2A/K-RAS2B/K-RAS4A/K-RAS4B/KI-RAS/KRAS1/KRAS2/NS3/RASK2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 3845 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |