missing translation for 'onlineSavingsMsg'
Learn More

Kinesin Mouse anti-Bovine, Human, Rat, Clone: H1, MilliporeSigma™

Catalog No. MAB1613MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1613MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1613MI Supplier MilliporeSigma Supplier No. MAB1613
Only null left

Mouse Monoclonal Antibody

Anti-Kinesin Antibody, heavy chain, non-neuronal KHC, clone H1 detects level of Kinesin and has been published and validated for use in ELISA, immunoprecipitation, RIA, western blot, immunocytochemistry, immunohistochemistry.

Specifications

Antigen Kinesin
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone H1
Conjugate Unconjugated
Formulation Liquid in 0.02MPhosphate buffer, 0.25MNaCl.pH 7.6 with 0.1% sodium azide.
Gene Accession No. NM_004521.1
Gene Symbols KIF5B, UKHC, KINH, KNS1, KNS
Host Species Mouse
Immunogen Bovine brain kinesin.
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only