missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KEAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Description
Anti-KEAP1 Antibody detects level of KEAP1 and has been published and validated for use in western blotting and immunocytochemistry.
Specifications
Specifications
| Antigen | KEAP1 |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Symbols | KEAP1; INRF2; KLHL19 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to the Kelch 3 region of human KEAP1. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only