missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDR/Flk-1/VEGFR2 Mouse anti-Human, Mouse, Clone: CH-11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05554
Description
Anti-KDR/Flk-1/VEGFR2 Antibody, clone CH-11 detects level of KDR/Flk-1/VEGFR2 and has been published and validated for use in ELISA, IC and western blotting.Specifications
| KDR/Flk-1/VEGFR2 | |
| Monoclonal | |
| Unconjugated | |
| NM_010612 | |
| Mouse | |
| Protein A chromatography | |
| Primary | |
| 2 years at 4°C | |
| IgG1 |
| ELISA, Immunocytochemistry, Western Blot | |
| CH-11 | |
| 20mM sodium phosphate, pH 7.6, 250mM NaCl with 0.1% sodium azide | |
| KDR, VEGFR-2, VEGFR2, VEGFR, CD309, FLK1 | |
| Recombinant protein corresponding to residues 1158-1345 of murine KDR/Flk-1 | |
| 100 μg | |
| Human, Mouse | |
| Purified |
For Research Use Only