missing translation for 'onlineSavingsMsg'
Learn More

JMJD2A Rabbit anti-Human, Clone: 8B7.1, MilliporeSigma™

Catalog No. MABE222MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE222MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE222MI Supplier MilliporeSigma Supplier No. MABE222
Only null left

Rabbit Monoclonal Antibody

This Anti-JMJD2A Antibody, clone 8B7.1 is validated for use in Western Blotting, immunohistochemistry for the detection of JMJD2A.

Specifications

Antigen JMJD2A
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8B7.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_055478
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human JMJD2A.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) KDM4A, JHDM3A, JMJD2, KIAA0677
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only