Learn More
JAK3 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 071487
Description
Detect JAK3 using this Anti-JAK3 Antibody validated for use in western blotting and immunoprecipitation.Specifications
| JAK3 | |
| Polyclonal | |
| Rabbit polyclonal IgG whole antiserum in buffer containing sodium azide. | |
| Rabbit | |
| 100 μL | |
| NM_000215.2 | |
| -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| JAK3; JAKL; LJAK; JAK3_HUMAN; L-JAK; JAK-3; EC 2.7.10.2 | |
| Synthetic peptide corresponding to amino acids 782-797 of mouse JAK3 (SDPTPGIPSPRDELC). | |
| Primary | |
| Human, Mouse | |
| Serum |
For Research Use Only