missing translation for 'onlineSavingsMsg'
Learn More

Interferon-α Mouse anti-Human, Clone: ST126, MilliporeSigma™

Catalog No. mab413mi
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB413MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB413MI Supplier MilliporeSigma Supplier No. MAB413
Only null left

Mouse Monoclonal Antibody

Anti-Interferon-α Antibody, clone ST126 detects level of Interferon-α and has been published and validated for use in NEUT.

Specifications

Antigen Interferon-α
Applications Neutralization
Classification Monoclonal
Clone ST126
Conjugate Unconjugated
Formulation Liquid, no preservatives.
Gene Accession No. NM_000605.2
Gene Symbols IFNA2, INFA2, MGC125764, IFNA, MGC125765
Host Species Mouse
Immunogen Human alpha-interferon
Quantity 500 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for up to 12 months in undiluted aliquots.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only