missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta-3 (CD61) Armenian Hamster anti-Mouse, Clone: HMb3-1, MilliporeSigma™
Armenian Hamster Monoclonal Antibody
Supplier: MilliporeSigma MAB2637
Description
Detect Integrin beta-3 (CD61) using this Anti-Integrin beta-3 (CD61) Antibody, clone HMb3-1 validated for use in flow cytometry, immunohistochemistry.Specifications
| Integrin beta-3 (CD61) | |
| Monoclonal | |
| Unconjugated | |
| NP_058060.2 | |
| Armenian Hamster | |
| Protein G purfied | |
| Primary | |
| 2-8°C for one year | |
| IgG |
| Flow Cytometry, Immunohistochemistry | |
| HMb3-1 | |
| Purified armenian hamster monoclonal IgG in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| CD61, GP3A, GPIIIa | |
| Recombinant protein corresponding to mouse Integrin beta-3. | |
| 100 μg | |
| Mouse | |
| Purified |
For Research Use Only