missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin α5Β1 Rat anti-Mouse, Clone: BMC5, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MAB2575
Description
Detect Integrin α5β1 using this Anti-Integrin α5β1 Antibody, clone BMC5 validated for use in flow cytometry, immunoprecipitation, immunohistochemistry.Specifications
| Integrin α5β1 | |
| Monoclonal | |
| Unconjugated | |
| NM_002205.2 | |
| Rat | |
| 100 μL | |
| Mouse | |
| Ascites |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| BMC5 | |
| Liquid ascites. | |
| ITGA5, VLA-5, CD49e, VLA5A, FNRA | |
| Mouse embryonal carcinoma P19 cells. | |
| Primary | |
| −20°C in undiluted aliquots for up to 12 months. | |
| IgG2c |
For Research Use Only