missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin α2Β1 Mouse anti-Canine, Human, Pig, Clone: BHA2.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1998
Description
Anti-Integrin α2β1 Antibody, clone BHA2.1 is an antibody against Integrin α2β1 for use in flow cytometry, immunohistochemistry, immunohistochemistry (paraffin) and immunoprecipitation.Specifications
| Integrin α2β1 | |
| Monoclonal | |
| Unconjugated | |
| NM_002203.3 | |
| Mouse | |
| 100 μg | |
| Canine, Human, Pig | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| BHA2.1 | |
| Purified immunoglobulin from Protein A chromatography. Presented in liquid in 0.02M PB pH 7.6, 0.25M NaCl, 0.1% sodium azide. | |
| ITGA2, CD49B, VLAA2, VLA-2, CD49b, BR, GPIa | |
| Protein A purfied | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 κ |
For Research Use Only