missing translation for 'onlineSavingsMsg'
Learn More

Integrin β1 Rabbit anti-Human, Mouse, Pig, Rat, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$684.23

Specifications

Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Target Species Human, Mouse, Pig, Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AB1952PMI
View Documents Promotion Details
N/A 100 μg
Each for $684.23
Only null left
 
AB1952MI
View Documents Promotion Details
N/A 100 μL
Each for $684.23
Only null left
 
Description

Description

Anti-Integrin beta1 Antibody, Cytosolic detects level of Integrin beta1 and has been published and validated for use in immunoprecipitation and western blotting.
Specifications

Specifications

Immunoprecipitation, Western Blot
Unconjugated
Human, Mouse, Pig, Rat
Polyclonal
Rabbit
Primary
Videos
SDS
Documents

Documents

For Research Use Only