Learn More
inhibitor of growth family, member 1, Mouse, Clone: 2F9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ING1.
Supplier: Abnova Corporation H00003621M05
Description
This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq
Sequence: DAKYQEILKELDECYERFSRETDGAQKRRMLHCVQRALIRSQELGDEKIQIVSQMVELVENRTRQVDSHVELFEAQQELGDTAGNSGKAGADRPKGEAAASpecifications
| inhibitor of growth family, member 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_198219.1 | |
| ING1 | |
| ING1 (NP_937862.1, 41 a.a. ∼ 140 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 50 μg | |
| Yes | |
| 3621 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgM κ |
| ELISA, Western Blot | |
| 2F9 | |
| Mouse monoclonal antibody raised against a partial recombinant ING1. | |
| ING1 | |
| p24ING1c/p33/p33ING1/p33ING1b/p47/p47ING1a | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |