missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID4 Mouse anti-Human, Mouse, Clone: DML07, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE164
Description
Use Anti-ID4 Antibody, clone DML07 (Mouse Monoclonal Antibody) validated in western blotting to detect ID4 also known as DNA-binding protein inhibitor ID-4, Class B basic helix-loop-helix protein 27, bHLHb27.Specifications
| ID4 | |
| Monoclonal | |
| Unconjugated | |
| NP_001537 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| 2-8°C for one year | |
| IgG2b κ |
| Western Blot | |
| DML07 | |
| Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| ID4 | |
| KLH-conjugated linear peptide corresponding to human ID4. | |
| 100 μg | |
| Human, Mouse | |
| Purified |
For Research Use Only