missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAD/DFF Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB16965
Description
Anti-ICAD/DFF Antibody, N-terminus (a.a. 2-21 of mouse ICAD) is an antibody against ICAD/DFF for use in western blotting.Specifications
| ICAD/DFF | |
| Polyclonal | |
| Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide. | |
| Rabbit | |
| 100 μg | |
| NM_004401.2; NM_213566.1 | |
| 2-8°C in undiluted aliquots for up to 12 months |
| Western Blot | |
| Unconjugated | |
| DFFA; DFF45; DFF-45; ICAD; DFF1 | |
| Polyclonal antibody raised against a peptide corresponding to amino acids 2-21 of mouse ICAD (Enari et al. 1998). | |
| Primary | |
| Mouse | |
| Purified |
For Research Use Only