missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1 Mouse anti-Human, Bovine, Green Plants, Clone: AE-4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05457
Description
Use Anti-Histone H1 Antibody, clone AE-4 (mouse monoclonal antibody) validated in ELISA, immunocytochemistry, western blotting to detect Histone H1 also known as H1 histone family member 4, Histone H1b, histone 1 H1e, histone cluster 1 H1e.Specifications
| Histone H1 | |
| Monoclonal | |
| Unconjugated | |
| NM_005322 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG2a |
| ELISA, Immunocytochemistry, Western Blot | |
| AE-4 | |
| Purified mouse monoclonal IgG2a in buffer containing PBS, pH 7.2, with 0.1% sodium azide. Frozen solution. | |
| HIST1H1E, H1F4, H1.4, dJ221C16.5, MGC116819 | |
| Nuclei from myeloid leukemia biopsy cells. Clone AE-4. | |
| 200 μg | |
| Bovine, Human, Plant | |
| Purified |
For Research Use Only