missing translation for 'onlineSavingsMsg'
Learn More

Herpes Virus 6 Mouse anti-Human, Clone: C3108-103, MilliporeSigma™

Catalog No. MAB8535MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8535MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8535MI Supplier MilliporeSigma Supplier No. MAB8535
Only null left

Mouse Monoclonal Antibody

This Anti-Herpes Virus 6 Antibody, 101kDa protein, clone C3108-103 is validated for use in immunofluoresence, western blot, immunohistochemistry for the detection of Herpes Virus 6.

Specifications

Antigen Herpes Virus 6
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone C3108-103
Conjugate Unconjugated
Formulation In 0.02M phoshate buffer, containing 0.25M NaCl and 0.1% Sodium Azide, pH 7.6.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only