Learn More
Anti-GT1b Monoclonal Antibody , TCI America™
Anti-GT1b Monoclonal Antibody, TCI America™
Supplier:  TCI America A27321VIAL
Catalog No. A2732VIAL
Only null left
Description
Description
Specifications
Specifications
GT1b
Monoclonal
Mouse
1 Vial
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
IgM κ
Flow Cytometry, Immunohistochemistry, ELISA, TLC-Immunostaining
GMR5
Purified ganglioside GT1b
Primary
Purified
Product Suggestions
Product Suggestions