missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-GM3 Monoclonal Antibody , TCI America™
Anti-GM3 Monoclonal Antibody, TCI America™
Supplier: TCI America A25821VIAL
Specifications
| GM3 | |
| Monoclonal | |
| Mouse | |
| Primary | |
| Purified |
| TLC-Immunostaining, Flow Cytometry, ELISA, Immunohistochemistry | |
| GMR6 | |
| 1 Vial | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
| IgM κ |