missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-GM2 Monoclonal Antibody , TCI America™
Anti-GM2 Monoclonal Antibody, TCI America™
Supplier: TCI America A25761VIAL
Specifications
| GM2 | |
| Monoclonal | |
| Mouse | |
| 1 Vial | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
| IgM κ |
| TLC-Immunostaining, ELISA, Immunohistochemistry, Flow Cytometry | |
| MK1-16 | |
| GM2(NeuAc)+Monophosphoryl lipid A+ Treharose imycolate emulsion | |
| Primary | |
| Purified |