missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Mouse anti-Eukaryote, Clone: D8, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5310
Description
Anti-Glutathione Antibody, clone D8 is an antibody against Glutathione for use in western blotting.Specifications
| Glutathione | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 100 μg | |
| Eukaryote | |
| Purified |
| Western Blot | |
| D8 | |
| Immunoglobulin purified by protein A affinity and presented as a liquid in PBS, pH 7.2 with 0.01% sodium azide. | |
| Glutathione-protein complexes. | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 6 months. | |
| IgG2a |
For Research Use Only