missing translation for 'onlineSavingsMsg'
Learn More

starch binding domain 1, Mouse, Clone: 4H4, Abnova™

Catalog No. 89001110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89001110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89001110 Supplier Abnova Corporation Supplier No. H00008987M03
Only null left

Mouse monoclonal antibody raised against a full-length recombinant GENX-3414.

Sequence: MGAVWSALLVGGGLAGALFVWLLRGGPGDTGKDGDAEQEKDAPLGGAAIPGGHQSGSSGLSPGPSGQELVTKPEHLQESNGHLISKTKDLGKLQAASWRLQNPSREVCDNSREHVPSGQFPDTEAPATSETSNSRSYSEVSRNESLESPMGEWGFQKGQEISAKAATCFAEKLPSSNLLKNRAKEEMSLSDLNSQDRVDHEEWEMVPRHSSWGDVGVGGSLKAPVLNLNQGMDNGRSTLVEARGQQVHGKMERVAVMPAGSQQVSVRFQVHYVTSTDVQFIAVTGDHECLGRWNTYIPLHYNKDGFWSHSIFLPADTVVEWKFVLVENGGVTRWEECSNRFLETGHEDKVVHAWWGIH

Specifications

Antigen starch binding domain 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4H4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant GENX-3414.
Formulation PBS with no preservative; pH 7.4
Gene STBD1
Gene Accession No. BC022301
Gene Alias FLJ41801/GENEX3414/GENX-3414
Gene Symbols STBD1
Host Species Mouse
Immunogen GENX-3414 (AAH22301, 1 a.a. ∼ 358 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8987
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less