missing translation for 'onlineSavingsMsg'
Learn More

Gemin3 Mouse anti-Human, Clone: 12H12, MilliporeSigma™

Catalog No. 051533MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
051533MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 051533MI Supplier MilliporeSigma Supplier No. 051533
Only null left

Mouse Monoclonal Antibody

Anti-Gemin3 Antibody, clone 12H12 is a Mouse Monoclonal Antibody for detection of Gemin3 also known as Component of gems 3, DEAD-box protein DP103 and has been validated in western blotting.

Specifications

Antigen Gemin3
Applications Western Blot
Classification Monoclonal
Clone 12H12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgGκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_009135.4
Gene Symbols DDX20, DP103, GEMIN3, Gemin-3
Host Species Mouse
Immunogen Recombinant protein corresponding to human Gemin-3.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only