missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gC1qR Mouse anti-Human, Clone: 74.5.2, MilliporeSigma™
Description
Anti-gC1qR Antibody, clone 74.5.2 detects level of gC1qR and has been published and validated for use in ELISA, flow cytometry, immunofluorescence and western blotting.
Specifications
Specifications
| Antigen | gC1qR |
| Applications | ELISA, Flow Cytometry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 74.5.2 |
| Conjugate | Unconjugated |
| Formulation | mouse ascites with 30% glycerol |
| Gene Accession No. | NM_001212 |
| Gene Symbols | C1QBP, C1qBP, p33, gC1Q-R, GC1QBP, gC1qR, GC1Q-R, HABP1, SF2P32, SF2p32, p32 |
| Host Species | Mouse |
| Immunogen | GST fusion protein corresponding to residues 74-282 of the mature form of human gC1qR. The epitope has been mapped to residues 204-218 |
| Show More |
For Research Use Only