missing translation for 'onlineSavingsMsg'
Learn More

GBP5 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABS381MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS381MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS381MI Supplier MilliporeSigma Supplier No. ABS381
Only null left

Rabbit Polyclonal Antibody

Detect GTP binding protein using this rabbit polyclonal antibody, Anti-GBP5 Antibody validated for use in western blotting.

Specifications

Antigen GBP5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.
Gene Symbols GBP5
Host Species Rabbit
Immunogen Linear peptide corresponding to human GBP5.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001127958
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only