Learn More
Anti-Gb3 Monoclonal Antibody Biotin Conjugate , TCI America™
Anti-Gb3 Monoclonal Antibody Biotin Conjugate, TCI America™
Supplier:  TCI America A28221VIAL
Catalog No. A2822VIAL
Only null left
Description
Description
Specifications
Specifications
Gb3
Monoclonal
Mouse
1 Vial
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
IgM κ
TLC-Immunostaining, ELISA, Immunohistochemistry
BGR23
Purified glycolipid adsorbed to Salmonella Minnesota:Gb3
Primary
Purified
Product Suggestions
Product Suggestions