missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH Rabbit anti-Human, Mouse, Rat, Bovine, Guinea Pig, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABS16
Description
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody is popular affinity purified polyclonal antibody. This antibody is supported by peer reviewed publications and reliably detects Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) is validated for use in western blotting.Specifications
| GAPDH | |
| Polyclonal | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Bovine, Guinea Pig, Human, Mouse, Rat |
| Western Blot | |
| Unconjugated | |
| GAPDH; G3PDH; GAPD; G3PD | |
| GST-tagged recombinant protein corresponding to the C-terminus of human GAPDH. | |
| 100 μg | |
| NP_002037 | |
| 2-8°C for 12 months |
For Research Use Only