missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-GalNAc-GD1a Monoclonal Antibody , TCI America™
Anti-GalNAc-GD1a Monoclonal Antibody, TCI America™
Supplier: TCI America A27011VIAL
Specifications
| GalNAc-GD1a | |
| Monoclonal | |
| Mouse | |
| 1 Vial | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
| IgM κ |
| Immunohistochemistry, ELISA, Western Blot, TLC-Immunostaining | |
| 2A3D2 | |
| Salmonella minnesota strain R595 coated with ganglioside mixture from human hepatocellular carcinoma cells, PLC/PRF/5 | |
| Primary | |
| Purified |