missing translation for 'onlineSavingsMsg'
Learn More

GABPA, Mouse, Clone: 5C8, Abnova™

Catalog No. 89022255
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89022255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89022255 Supplier Abnova Corporation Supplier No. H00002551M07
Only null left

Mouse monoclonal antibody raised against a partial recombinant GABPA.

This gene encodes one of three GA-binding protein transcription factor subunits which functions as a DNA-binding subunit. Since this subunit shares identity with a subunit encoding the nuclear respiratory factor 2 gene, it is likely involved in activation of cytochrome oxidase expression and nuclear control of mitochondrial function. This subunit also shares identity with a subunit constituting the transcription factor E4TF1, responsible for expression of the adenovirus E4 gene. Because of its chromosomal localization and ability to form heterodimers with other polypeptides, this gene may play a role in the Down Syndrome phenotype. [provided by RefSeq

Sequence: MTKREAEELIEIEIDGTEKAECTEESIVEQTYAPAECVSQAIDINEPIGNLKKLLEPRLQCSLDAHEICLQDIQLDPERSLFDQGVKTDGTVQLSVQVIS

Specifications

Antigen GABPA
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 5C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GABPA.
Formulation PBS with no preservative; pH 7.4
Gene GABPA
Gene Accession No. NM_002040
Gene Alias E4TF1-60/E4TF1A/NFT2/NRF2/NRF2A
Gene Symbols GABPA
Host Species Mouse
Immunogen GABPA (NP_002031, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2551
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less