missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA B Receptor R2 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5848
Description
This Anti-GABA B Receptor R2 Antibody, a.a. 42-54 rat is validated for use in western blotting for the detection of GABA B Receptor R2.Specifications
| GABA B Receptor R2 | |
| Polyclonal | |
| Purified immunoglobulin. Liquid. Buffer = PBS containing 0.02% sodium azide. | |
| Rabbit | |
| 100 μg | |
| NM_005458.5 | |
| −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| GABBR2; HRIHFB2099; OTTHUMP00000063797; GPRC3B; GPR51; HG20; Gb2; FLJ36928; GABA-B-R2; GABABR2 | |
| peptide residues 42-54 of the rat 105 kDa GABA(b)R2 protein | |
| Primary | |
| Human, Rat | |
| Purified |
For Research Use Only