missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA(A)R, Alpha1 Protein Mouse anti-Mouse, Rat, Clone: N95/35, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN489
Description
This Anti-GABA(A)R, Alpha1 Protein Antibody, clone N95/35 is validated for use in Western Blotting, immunohistochemistry for the detection of GABA(A)R, Alpha1 Protein, clone N95/35.Specifications
| GABA(A)R | |
| Monoclonal | |
| Unconjugated | |
| NP_034380 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| 2-8°C for one year | |
| IgG2a |
| Immunohistochemistry, Western Blot | |
| N95/35 | |
| Purified mouse monoclonal IgG2a in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Gabra1, Gabra-1 | |
| Recombinant protein corresponding to the cytoplasmic domain of mouse GABA(A)R, Alpha1 Protein. | |
| 100 μg | |
| Mouse, Rat | |
| Purified |
For Research Use Only