missing translation for 'onlineSavingsMsg'
Learn More

GΒ5 Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™

Catalog No. 071208mi
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
071208MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 071208MI Supplier MilliporeSigma Supplier No. 071208
Only null left

Rabbit Polyclonal Antibody

This Anti-Gβ5 Antibody is validated for use in western blotting for the detection of Gβ5.

Specifications

Antigen Gβ5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in phosphate buffered saline with 0.08% sodium azide.
Gene Symbols GNB5; FLJ37457; FLJ43714; Gbeta5; GB5
Host Species Rabbit
Immunogen Unique peptide sequence derived from the D1 blade of the Gβ5 protein.
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006578.3
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only