missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMC-7 B-Cell Lymphocyte Marker Mouse anti-Human, FITC, Clone: FMC-7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1217F
Description
This Anti-FMC-7 B-Cell Lymphocyte Marker Antibody, clone FMC-7, FITC conjugated is validated for use in Flow Cytometry (FC) for the detection of FMC-7 B-Cell Lymphocyte Marker.Specifications
| Fmc-7 (B-Cell Lymphocyte Marker) | |
| Monoclonal | |
| FITC | |
| FMC-7, FMC7 | |
| The human B-lymphoblastoid cell line, HRIK. | |
| Primary | |
| 2-8°C for 6 months. Do not freeze. Protect from light. |
| Flow Cytometry | |
| FMC-7 | |
| The antibody is supplied in 1.0mL 0.1M Tris-HCl, 0.5M NaCl, 10mM glycine, pH 8.0, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry. | |
| Mouse | |
| 1 Unit | |
| Human | |
| IgM |
For Research Use Only