missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fmc-7 (B-Cell Lymphocyte Marker) Mouse anti-Human, Clone: Fmc-7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1217I
Description
This Anti-Fmc-7 (B-Cell Lymphocyte Marker) antibody is validated for use in western blot, FC for the detection of Fmc-7 (B-Cell Lymphocyte Marker).Specifications
| Fmc-7 (B-Cell Lymphocyte Marker) | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Ammonium Sulfate | |
| Primary | |
| 2-8°C for one year | |
| IgM κ |
| Flow Cytometry, Western Blot | |
| Fmc-7 | |
| Purified mouse monoclonal IgMκ in buffer containing 0.1M Tris, 10mM Glycine, 0.5M NaCl, and 0.1% Sodium Azide. | |
| HRIK [human B-lymphoblastoid cell] corresponding to human Fmc-7 (B-Cell Lymphocyte Marker). | |
| 100 Tests | |
| Human | |
| Purified |
For Research Use Only