missing translation for 'onlineSavingsMsg'
Learn More

filamin C, gamma (actin binding protein 280), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004126
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89004126 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89004126 Supplier Abnova Corporation Supplier No. H00002318A01
Only null left

Mouse polyclonal antibody raised against a partial recombinant FLNC.

This gene encodes one of three related filamin genes, specifically gamma filamin. These filamin proteins crosslink actin filaments into orthogonal networks in cortical cytoplasm and participate in the anchoring of membrane proteins for the actin cytoskeleton. Three functional domains exist in filamin: an N-terminal filamentous actin-binding domain, a C-terminal self-association domain, and a membrane glycoprotein-binding domain. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: SSIPKFSSDASKVVTRGPGLSQAFVGQKNSFTVDCSKAGTNMMMVGVHGPKTPCEEVYVKHMGNRVYNVTYTVKEKGDYILIVKWGDESVPGSPFKVKVP

Specifications

Antigen filamin C, gamma (actin binding protein 280)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant FLNC.
Formulation 50% glycerol
Gene FLNC
Gene Accession No. NM_001458
Gene Alias ABP-280/ABP280A/ABPA/ABPL/FLJ10186/FLN2
Gene Symbols FLNC
Host Species Mouse
Immunogen FLNC (NP_001449.3, 2606 a.a. ∼ 2705 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2318
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less