missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGF-like, fibronectin type III and laminin G domains, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FLJ39155 protein.
Supplier: Abnova Corporation H00133584B01P
Description
Sequence: MRFKTTAKDGLLLWRGDSPMRPNSDFISLGLRDGALVFSYNLGSGVASIMVNGSFNDGRWHRVKAVRDGQSGKITVDDYGARTGKSPGMMRQLNINGALYVGGMKEIALHTNRQYMRGLVGCISHFTLSTDYHISLVEDAVDGKNINTCGAKSpecifications
| EGF-like, fibronectin type III and laminin G domains | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human FLJ39155 protein. | |
| EGFLAM | |
| AGRINL/AGRNL/FLJ39155 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_182801.1 | |
| EGFLAM | |
| FLJ39155 (NP_877953.1, 1 a.a. ∼ 152 a.a) full-length human protein. | |
| 50 μg | |
| Cell Adhesion | |
| 133584 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |