missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ10986, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FLJ10986.
Supplier: Abnova Corporation H00055277A01
Description
Sequence: MWLDHRAVSQVNRINETKHSVLQYVGGVMSVEMQAPKLLWLKENLREICWDKAGHFFDLPDFLSWKATGVTARSLCSLVCKWTYSAEKGWDDSFWKMIGSpecifications
| FLJ10986 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant FLJ10986. | |
| FGGY | |
| FLJ10986 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_018291 | |
| FGGY | |
| FLJ10986 (NP_060761, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 55277 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |