missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF23 Mouse anti-Human, Clone: 278-2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS460
Description
Anti-FGF23 Antibody, clone 278-2 is a mouse monoclonal antibody and validated for use in western blotting.Specifications
| FGF23 | |
| Monoclonal | |
| Unconjugated | |
| NP_065689 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| 2-8°C for one year | |
| IgG1 κ |
| Western Blot | |
| 278-2 | |
| Purified mouse monoclonal IgG1κ in buffer containing PBS with 0.05% sodium azide. | |
| FGF23, HYPF | |
| Corresponding to human FGF23. | |
| 100 μg | |
| Human | |
| Purified |
For Research Use Only