missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fas ligand (TNF superfamily, member 6), Mouse, Clone: 2G9-G8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FASLG.
Supplier: Abnova Corporation H00000356M02
Description
The protein encoded by this gene is the ligand for FAS. Both are transmembrane proteins. Interaction of FAS with this ligand is critical in triggering apoptosis of some types of cells such as lymphocytes. Defects in this gene may be related to some cases of systemic lupus erythematosus (SLE). [provided by RefSeq
Sequence: MQQPFNYPYPQIYWVDSSASSPWAPPGTVLPCPTSVPRRPGQRRPPPPPPPPPLPPPPPPPPLPPLPLPPLKKRGNHSTGLCLLVMFFMVLVALVGLGLGMFQLFHLQKELAELRESTSQMHTASSLEKQIGHPSPPPEKKELRKVAHLTGKSNSRSMPLEWEDTYGIVLLSGVKYKKGGLVINETGLYFVYSKVYFRGQSCNNLPLSHKVYMRNSKYPQDLVMMEGKMMSYCTTGQMWARSSYLGAVFNLTSADHLYVNVSELSLVNFEESQTFFGLYKLSpecifications
| Fas ligand (TNF superfamily, member 6) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC017502 | |
| FASLG | |
| FASLG (AAH17502.1, 1 a.a. ∼ 281 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Apoptosis | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, In situ PLA, Western Blot | |
| 2G9-G8 | |
| Mouse monoclonal antibody raised against a full length recombinant FASLG. | |
| FASLG | |
| APT1LG1/CD178/CD95L/FASL/TNFSF6 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 356 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |