missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAK Mouse anti-Human, Mouse, Rat, Clone: 4.47, MilliporeSigma™
Description
Anti-FAK Antibody, clone 4.47 is an antibody against FAK for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
Specifications
Specifications
| Antigen | FAK |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 4.47 |
| Conjugate | Unconjugated |
| Formulation | Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.1mM EDTA, 0.05% sodium azide before the addition of glycerol to 30% |
| Gene Accession No. | NM_005607 / NM_153831 |
| Gene Symbols | PTK2, pp125FAK, FAK, FADK, FAK1 |
| Host Species | Mouse |
| Immunogen | GST fusion protein corresponding to residues 1-423 of human FAK (focal adhesion kinase); specific for p125FAK |
| Show More |
For Research Use Only