missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VIII Mouse anti-Human, Clone: GMA-012, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05871
Description
Detect Factor VIII using this Anti-Factor VIII Antibody, clone GMA-012 validated for use in ELISA and western blotting.Specifications
| Factor VIII | |
| Monoclonal | |
| Unconjugated | |
| F8, F8C, OTTHUMP00000061446, FVIII, HEMA, AHF, DXS1253E, F8B | |
| Purified human Factor VIII | |
| 100 μg | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| GMA-012 | |
| NM_000132 / NM_019863 | |
| Mouse | |
| DEAE purified | |
| Primary | |
| −20°C for up to 24 months. | |
| IgG1 |
For Research Use Only