Learn More
FABP6, Mouse, Clone: 4A4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FABP6.
Supplier: Abnova Corporation H00002172M01
Description
This gene encodes the ileal fatty acid binding protein. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP6 and FABP1 (the liver fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. Transcript variants generated by alternate transcription promoters and/or alternate splicing have been found for this gene. [provided by RefSeq
Sequence: MAFTGKFEMESEKNYDEFMKLLGISSDVIEKAHNFKIVTEVQQDGQDFTWSQHYYGGHTMTNKFTVGKESNIQTMGGKTFKATVQMEGGKLVVNFPNYHQTSEIVGDKLVEVSTIGGVTYERVSKRLASpecifications
| FABP6 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC022489 | |
| FABP6 | |
| FABP6 (AAH22489, 1 a.a. ∼ 128 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4A4 | |
| Mouse monoclonal antibody raised against a full-length recombinant FABP6. | |
| FABP6 | |
| I-15P/I-BABP/I-BALB/I-BAP/ILBP/ILBP3/ILLBP | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2172 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |