missing translation for 'onlineSavingsMsg'
Learn More

F-actin-capping protein subunit beta Rabbit anti-Human, Mouse, Rat, Canine, Primate, Bovine, Polyclonal, MilliporeSigma™

Catalog No. AB6017MI
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB6017MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB6017MI Supplier MilliporeSigma Supplier No. AB6017
Only null left

Rabbit Polyclonal Antibody

Detect F-actin-capping protein subunit beta using this Anti-F-actin-capping protein subunit beta Antibody validated for use in western blotting and immunocytochemistry.

Specifications

Antigen F-actin-capping protein subunit beta
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine, pH 7.4, 150mM NaCl, with 0.05% sodium azide.
Gene Symbols CAPZ; CAPPB; CAPB; Cap Z; CapZ beta; CAZB
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human F-actin-capping protein subunit beta of the C-terminus of isoform 2.
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_004921
Target Species Bovine, Canine, Human, Mouse, Primate, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only