missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enhancer of rudimentary homolog (Drosophila), Mouse, Clone: 4A10, Abnova™
Description
Sequence: MSHTILLVQPTKRPEGRTYADYESVNECMEGVCKMYEEHLKRMNPNSPSITYDISQLFDFIDDLADLSCLVYRADTQTYQPYNKDWIKEKIYVLLRRQAQQAGK
Specifications
Specifications
| Antigen | enhancer of rudimentary homolog (Drosophila) |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 4A10 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant ERH. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ERH |
| Gene Accession No. | BC014301 |
| Gene Alias | DROER/FLJ27340 |
| Show More |