missing translation for 'onlineSavingsMsg'
Learn More

EPH receptor A7, Mouse, Clone: 3D11, Abnova™

Catalog No. 89001449
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89001449 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89001449 Supplier Abnova Corporation Supplier No. H00002045M02A

Mouse monoclonal antibody raised against a full-length recombinant EPHA7.

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. [provided by RefSeq

Sequence: MVFQTRYPSWIILCYIWLLRFAHTGEAQAAKEVLLLDSKAQQTELEWISSPPNGWEEISGLDENYTPIRTYQVCQVMEPNQNNWLRTNWISKGNAQRIFVELKFTLRDCNSLPGVLGTCKETFNLYYYETDYDTGRNVRENLYVKIDTIAADESFTQGDLGERKMKLNTEVREIGPLSKKGFYLAFQDVGACIALVSVKVYYKKCWSIIENLAIFPDTVTGSEFSSLVEVRGTCVSSAEEEAENAPRMHCSAEGEWLVPIGKCICKAGYQQKGDTCECK

Specifications

Antigen EPH receptor A7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant EPHA7.
Formulation ascites with no preservative
Gene EPHA7
Gene Accession No. BC027940
Gene Alias EHK3/HEK11
Gene Symbols EPHA7
Host Species Mouse
Immunogen EPHA7 (AAH27940.1, 1 a.a. ∼ 279 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2045
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less