Learn More
Anti-Endo-M Polyclonal Antibody Biotin Conjugate , TCI America™
Anti-Endo-M Polyclonal Antibody Biotin Conjugate, TCI America™
Supplier:  TCI America A29591VIAL
Catalog No. A2959VIAL
Only null left
Description
Description
Specifications
Specifications
Endo-M
Polyclonal
endo-β-N-Acetylglucosaminidase (Endo-M) from Mucor hiemalis
Primary
Purified
ELISA, Western Blot
Rabbit
1 vial
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
IgG
Product Suggestions
Product Suggestions