Learn More
elongation protein 4 homolog (S. cerevisiae) (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ELP4.
Supplier: Abnova Corporation H00026610A01
Description
This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined. [provided by RefSeq
Sequence: YFLAEGIVNGHTLLVASAKEDPANILQELPAPLLDDKCKKEFDEDVYNHKTPESNIKMKIAWRYQLLPKMEIGPVSSSRFGHYYDASKRMPQELIEASNSpecifications
| elongation protein 4 homolog (S. cerevisiae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ELP4. | |
| ELP4 | |
| C11orf19/FLJ20498/PAX6NEB/PAXNEB/dJ68P15A.1 | |
| Mouse | |
| 50 μL | |
| Kinases | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_019040 | |
| ELP4 | |
| ELP4 (NP_061913, 101 a.a. ∼ 199 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 26610 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |