missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF3, Mouse, Clone: 2G9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ELF3.
Supplier: Abnova Corporation H00001999M05
Description
Sequence: EGKKSKHAPRGTHLWEFIRDILIHPELNEGLMKWENRHEGVFKFLRSEAVAQLWGQKKKNSNMTYEKLSRAMRYYYKREILERVDGRRLVYKFGKNSSGWKEEEVLQSRNSpecifications
| ELF3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC003569 | |
| ELF3 | |
| ELF3 (AAH03569, 262 a.a. ∼ 371 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2G9 | |
| Mouse monoclonal antibody raised against a partial recombinant ELF3. | |
| ELF3 | |
| EPR-1/ERT/ESE-1/ESX | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1999 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |